missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B-Myb Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-53779
This item is not returnable.
View return policy
Description
B-Myb Polyclonal specifically detects B-Myb in Rat samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Peptide with sequence C-QEKARQLLGRLKPSH corresponding to C-Terminus according to NP_002457.1. | |
| RUO | |
| IgG |
| 0.1 mg | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction