Learn More
Fryka™ B 30 Cold Box
Product Code. 10270292
Change view
Click to view available options
Capacity (Metric):
30 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10270292 30 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10270292 Supplier Fryka™ Supplier No. 050B3020
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
Compact and space saving. Fryka B 30 Cold Box designed as desktop appliances and are used for cooling and freezing for all research and industry laboratories.
  • Insulating glass plane in door
  • All stainless steel casing
  • Refrigeration unit: low noise, fully hermetically sealed, air-cooled, low maintenance
  • Interior free of fixtures
  • Electronic temperature control with LED display
  • Lockable door
  • Bushing (diameter 10 mm), e.g. for temperature monitoring with external sensors
  • Ram protection ring going over and under
  • Stackable unit, max. 3 appliances
Specifications
Type Cold Box
Capacity (Metric) 30 L
Temperature Range 12°C to 30°C
For Use With (Application) Research and industry laboratories
Model B30
Dimensions (D x W x H) Interior 35 x 36 x 23 cm
Depth (Metric) Exterior 70 cm
Height (Metric) Exterior 46 cm
Width (Metric) Exterior 53 cm
Electrical Requirements 230 V 50 Hz