Learn More
Azlon™ LDPE Wide Neck Wash Bottles
Product Code. p-8001811
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Closure Color:
Assorted
Blue
Green
Red
White
Yellow
Unit Size:
Each
Pack of 5
Product Code. 12039847
Brand: Azlon WGW234P
Only null left
Add to basket
Edge
Add to basket
Use these translucent plastic bottles for a variety of liquid storage and sampling applications. Azlon™ LDPE Wide Neck Wash Bottles come with color-coded one-piece PP closures that maintain their shape in use.
Use these translucent plastic bottles for a variety of liquid storage and sampling applications. AzlonTM LDPE Wide Neck Wash Bottles come with color-coded one-piece PP closures that maintain their shape in use.
  • Square shoulder wash bottles featuring a wide neck for easy spill free filling
  • Fitted with color coded one-piece PP closures that maintain their shape in use
  • Closures feature precision jet-tip aperture for fine control
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Translucent
Includes Blue Polypropylene closure
Material LDPE
Quantity 5/PK
Temperature Range (Metric) –50 to 80°C
Closure Color Blue
Closure Included Yes
Graduated No
Mouth Wide
Shape Round
Height (Metric) 140 mm
Product Type Wash Bottle
Width (English) 2.36 in.
Height (English) 5.51 in.
Closure Material Polypropylene
Diameter (Metric) 60 mm
Closure Type Cap
Through the Closure Dispensing Yes
Autoclavable Not Autoclavable