Learn More
Azlon™ LDPE Narrow Neck Round Bottles
Click to view available options
Capacity (Metric):
30 mL
60 mL
100 mL
125 mL
150 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 10
Pack of 5
Description
Use these Translucent, flexible plastic bottles for a wide variety of liquid storage and sampling applications. Azlon™ LDPE Narrow Neck Round Bottles are heavy duty and durable.
- Translucent, flexible LDPE bottles with polypropylene screw caps
- Suitable for food contact applications in accordance with EU and USA regulations
- Batch certificate available to download online
- Bulk packs contain an equal number of bottles and caps, packed separately
- Good general chemical resistance
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Color | Translucent |
| Material | LDPE |
| No. per Pack | 420 |
| Packaging | Bulk |
| Quantity | 420/Pk. |
| Mouth | Narrow |
| Shape | Round |
| Height (Metric) | 89 mm |
| Diameter (Metric) | 50 mm |
| Show More |