missing translation for 'onlineSavingsMsg'
Learn More

Azelastine Hydrochloride EP Impurity E (as (Z)-Isomer), Mikromol™

Product Code. 30051323
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
30051323 25 mg 25mg
30049789 100 mg 100mg
2 options
This item is not returnable. View return policy

Product Code. 30051323

Brand: Mikromol MM0362.060025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Azelastine Hydrochloride EP Impurity E (as (Z)-Isomer)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (Z)-3-(4-Chlorobenzylidene)isobenzofuran-1(3H)-one
CAS 105279-16-1
Analyte or Component Names (Z)-3-(4-Chlorobenzylidene)isobenzofuran-1(3H)-one
Shipping Condition Room Temperature
Molecular Formula C15 H9 Cl O2
Certifications/Compliance ISO 17025
Synonym Azelastine Hydrochloride Imp. E (EP) Z-Isomer, Azelastine Imp. E (EP) Z-Isomer, (Z)-3-(4-Chlorobenzylidene)isobenzofuran-1(3H)-one, Azelastine Hydrochloride Impurity E as Z-Isomer, Azelastine Impurity E as Z-Isomer
InChI Formula InChI=1S/C15H9ClO2/c16-11-7-5-10(6-8-11)9-14-12-3-1-2-4-13(12)15(17)18-14/h1-9H/b14-9-
SMILES Clc1ccc(\C=C\2/OC(=O)c3ccccc23)cc1
IUPAC Name (3Z)-3-[(4-chlorophenyl)methylidene]-2-benzofuran-1-one
Molecular Weight (g/mol) 256.68
Quantity 25 mg
Formula Weight 256.0291 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.