Description
- Available in sterile or nonsterile forms
- e-Beam irradiated to prevent product degradation
- Maxymum recovery tips allows complete sample retrieval and greater tip-to-tip precision
- Graduation marks at 110 μL and 500 μL, allows visual confirmation of pipettor draw volume
- Volume: 1000 μL
- Color: Blue
Order Info
- Bulk packed: 500 Tips/Pack. 10 Packs/Cs.
- Racked: 96 Tips/Rack, 12 Racks/Pack, 3 Packs/Cs.
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Sterility | Sterile |
| Color | Blue |
| Format | Rack |
| Tip Style | Low Retention |
| Packaging | Racked |
| For Use With (Equipment) | Eppendorf |
| Graduated | Yes |
| Graduations | 110 and 500μL |