missing translation for 'onlineSavingsMsg'
Learn More

Axin-2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

218.00€

Specifications

Antigen Axin-2
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL, Immunofluorescence 20 μg/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18593692
View Documents
Novus Biologicals
NBP1-76526-0.025mg
0.025 mg
218.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Axin-2 Polyclonal specifically detects Axin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Axin-2
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL, Immunofluorescence 20 μg/mL
Polyclonal
Rabbit
Signal Transduction, Wnt Signaling Pathway
PBS with 0.02% Sodium Azide
Axil, axin 2, axin-2, Axin-like protein, Axis inhibition protein 2, Conductin, DKFZp781B0869, MGC10366, MGC126582
AXIN2
IgG
Affinity Purified
Axin-2 antibody is not predicted to cross-react with AXIN1. At least four isoforms of AXIN2 are known to exist; this antibody will recognize all four.
1 mg/mL
Western Blot
Unconjugated
RUO
Human
AAF22799
8313
Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human AXIN2. The immunogen is located within amino acids 780 - 830 of AXIN2. Amino Acid Squence: KEQLSKKGNYRYYFKKASDE
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
85, 93 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.