missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axin-2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18241AF700
This item is not returnable.
View return policy
Description
Axin-2 Polyclonal antibody specifically detects Axin-2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Axin-2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| Axil, axin 2, axin-2, Axin-like protein, Axis inhibition protein 2, Conductin, DKFZp781B0869, MGC10366, MGC126582 | |
| Synthetic peptide from the mid-protein of Human Axin-2 (aa. 200-300) | |
| 0.1 mL | |
| Signal Transduction, Wnt Signaling Pathway | |
| 8313 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction