missing translation for 'onlineSavingsMsg'
Learn More

Chorionic Gonadotropin alpha/beta (hCG) Antibody (HCGab/52), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-45334-0.02MG

 View more versions of this product

Product Code. 18110279

  • 290.00€ / 0.02mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ensure accurate, reproducible results in Immunohistochemistry (Paraffin)

Chorionic Gonadotropin alpha/beta (hCG) Monoclonal specifically detects Chorionic Gonadotropin alpha/beta (hCG) in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

hCG holo C2
Monoclonal
0.2mg/mL
Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
choriogonadotropin subunit beta, CGB3, CGB5, CGB7, CGB8, chorionic gonadotropin, beta polypeptide, hCGB
Purified hCG protein
0.02 mg
Primary
This MAb is a very special because it reacts ONLY with the intact-HCG and not with either free alpha- or free beta-chain of HCG. HCG is a glycoprotein and is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and beta subunits. The alpha subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). HCG is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG MAb detects cells and tumors of trophoblastic origin such as choriocarcinoma. Large cell carcinoma and adenocarcinoma of the lung demonstrate anti-hCG positivity in 90% and 60% of cases respectively. 20% of lung squamous cell carcinomas are positive. HCG expression by non-trophoblastic tumors may indicate aggressive behavior.
Store at 4C.
IgG1 κ
Immunohistochemistry (Paraffin)
HCGab/52
Unconjugated
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Mouse
Protein A or G purified
RUO
1082
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only