missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza A H5N1 Hemagglutinin Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
622.00€
Specifications
| Antigen | Avian Influenza A H5N1 Hemagglutinin |
|---|---|
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Form | Purified |
Description
Avian Influenza A H5N1 Hemagglutinin Polyclonal antibody specifically detects Avian Influenza A H5N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISASpecifications
| Avian Influenza A H5N1 Hemagglutinin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Virus | |
| Antibody was raised against a synthetic peptide corresponding to 15 amino acids at the amino terminus of the avian influenza hemagglutinin protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 90 - 140 of Avian Influenza Hemagglutinin. Amino Acid Squence: CYPGDFNDYEELKHL | |
| Primary |
| Western Blot, ELISA | |
| mFluor Violet 450 SE | |
| Rabbit | |
| Infections (Virus Bacteria and Parasites) | |
| Harvey rat sarcoma viral oncogene homolog | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title