missing translation for 'onlineSavingsMsg'
Learn More

AUTS2 Antibody, Novus Biologicals™

Product Code. 18268240 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18268240 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18268240 Supplier Novus Biologicals Supplier No. NBP132784

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AUTS2 Polyclonal antibody specifically detects AUTS2 in Human, Mouse, Zebrafish samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AUTS2
Applications Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS, 20% Glycerol (pH7) with 0.025% Proclin 300
Gene Accession No. Q8WXX7
Gene Alias autism susceptibility candidate 2, autism susceptibility gene 2 protein, autism-related protein 1, KIAA0442FBRSL2, MGC13140
Gene Symbols AUTS2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human AUTS2. The exact sequence is proprietary.
Molecular Weight of Antigen 139 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26053
Test Specificity Human and Mouse. Not yet tested in any other species.
Target Species Human, Mouse, Zebrafish
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.