Learn More
- Home
- Products
- Pipette Products
- Pipette Tips
- Robotic Pipette Tips
- Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms
Product Code. 10729601
| Product Code. | Volume (Metric) | Sterility | Tip Style | Quantity | Unit Size |
|---|---|---|---|---|---|
| 10729601 | 1000 μL | Non-sterile | Automation | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10181471 | 10 μL | Sterile | Automation | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10338890 | 20 μL | Sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10013660 | 20 μL | Non-sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10623482 | 20 μL | Non-sterile | Conductive | 96 Tips/Tray, 50 Trays/Case | Pack of 4800 |
| 10461093 | 20 μL | Sterile | Automation | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10000481 | 20 μL | Non-sterile | Automation | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10697831 | 50 μL | Sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10020491 | 50 μL | Sterile | Automation | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10115430 | 50 μL | Sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10740981 | 50 μL | Non-sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10633482 | 50 μL | Non-sterile | Conductive | 96 Tips/Tray, 50 Trays/Case | Pack of 4800 |
| 10663242 | 50 μL | Sterile | Automation | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10673242 | 50 μL | Non-sterile | Automation | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10125430 | 175 μL | Sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10562162 | 175 μL | Sterile | Conductive, Wide Bore | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10141571 | 175 μL | Sterile | Automation | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 11510145 | 175 μL | Non-sterile | Conductive | 96 Tips/Tray, 50 Trays/Case | Pack of 4800 |
| 10439970 | 200 μL | Sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10561972 | 200 μL | Sterile | Conductive, Wide Bore | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10648221 | 200 μL | Non-sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10772563 | 200 μL | Non-sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10102821 | 200 μL | Non-sterile | Conductive | 96 Tips/Tray, 50 Trays/Case | Pack of 4800 |
| 10060511 | 200 μL | Sterile | Automation | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10759221 | 200 μL | Non-sterile | Automation | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10658221 | 1000 μL | Sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10702953 | 1000 μL | Sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 16446939 | 1000 μL | Non-sterile | Conductive | 96 Tips/Tray, 50 Trays/Case | Pack of 4800 |
| 10345981 | 1000 μL | Sterile | Automation | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10658411 | 1000 μL | Sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10231482 | 1000 μL | Sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10348980 | 1000 μL | Non-sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10572162 | 1000 μL | Non-sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10565051 | 1000 μL | Non-sterile | Conductive | 96 Tips/Tray, 50 Trays/Case | Pack of 4800 |
| 10306031 | 1000 μL | Sterile | Automation | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
| 10073620 | 10 μL | Sterile | Conductive | 96 Tips/Insert, 10 Inserts/Pk. | Pack of 960 |
Use promo code 28003
Log in to see stock.
BEST PRICE on Promo! Use promo code "28003" to get your promotional price.
Description
Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
- Available nonsterile with-or-without Thermo Scientific™ ART™ filter technology
- Available in filtered, low retention or extended length for increased volume to reach into deep well plates
- Lot-tested during production on actual workstation for which they are created
- Fully conductive providing increased pipetting performance
- Low percentage coefficient of variation (%CV)
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Rack Color | Gray |
| Autoclavable | Not Autoclavable |
| Certifications/Compliance | ISO 9001 and ISO 13485 |
| Clean Claims | Free of RNase, DNase, DNA, ATP, and endotoxin |
| Format | Reload Deck |
| Tip Style | Automation |
| Packaging | Blister Pack |
| Quantity | 96 Tips/Insert, 10 Inserts/Pk. |
| For Use With (Equipment) | Tecan Systems with LiHa Head |
| Fit for Purpose | Certified Quality |
| Show More Show Less |