Learn More
Abnova™ ATRX/CENXp FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
1475.00€ - 2430.00€
Specifications
Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
---|---|
For Use With (Application) | FISH-Ce |
Includes | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023375
|
Abnova™
FG0228.100UL |
100 uL |
1475.00€
100µL |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
16033375
|
Abnova™
FG0228.200UL |
200 uL |
2430.00€
200µL |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
Specifications
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
FISH-Ce | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |