missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATRIP, p Ser239 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-29537
This item is not returnable.
View return policy
Description
ATRIP Polyclonal specifically detects ATRIP in Human, Mouse samples. It is validated for Western Blot.
Specifications
| ATRIP (p Ser239) | |
| Polyclonal | |
| Western Blot 1:500 | |
| AGS1, ATM and Rad3-related-interacting protein, ATR interacting protein, ATR-interacting protein, DKFZp762J2115, FLJ12343, MGC20625, MGC21482, MGC26740 | |
| Rabbit | |
| 86 kDa | |
| 0.1 mL | |
| Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair | |
| 84126 | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| ATRIP | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser239 conjugated to KLH. Accession # Q8WXE1 | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Specific for the ∼86k ATRIP protein phosphorylated at Ser239. The antibody also recognizes a band at ∼32k. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction