Learn More
Atrazine, Dr. Ehrenstorfer
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Atrazine |
| CAS | 1912-24-9 |
| Analyte or Component Names | Atrazine |
| Shipping Condition | +20°C |
| Molecular Formula | C8 H14 Cl N5 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 1,3,5-Triazine-2,4-diamine, 6-chloro-N2-ethyl-N4-(1-methylethyl)-, 1,3,5-Triazine-2,4-diamine, 6-chloro-N-ethyl-N'-(1-methylethyl)- (9CI), s-Triazine, 2-chloro-4-(ethylamino)-6-(isopropylamino)- (8CI), 6-Chloro-N2-ethyl-N4-(1-methylethyl)-1,3,5-triazine-2,4-diamine, 1-Chloro-3-ethylamino-5-isopropylamino-2,4,6-triazine, 2-Chloro-4-(ethylamino)-6-(2-propylamino)-s-triazine, 2-Chloro-4-(ethylamino)-6-(isopropylamino)-s-triazine, 2-Chloro-4-(ethylamino)-6-(isopropylamino)triazine, 2-Chloro-4-ethylamineisopropylamine-s-triazine, 2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine, 2-Ethylamino-4-isopropylamino-6-chloro-1,3,5-triazine, 2-Ethylamino-4-isopropylamino-6-chloro-s-triazine, 6-Chloro-4-(ethylamino)-2-(isopropylamino)-s-triazine, 6-Chloro-N-Ethyl-N'-(1-methylethyl)-1,3,5-triazine-2,4-diamine, A 361, AAtrex, AAtrex 4L, AAtrex Nine-O, ATZ, Aatrax, Aatrex 4LC, Aatrex L, Akticon, Aktikon, Aktikon PK, Aktinit A, Aktinit PK, Argezin, Atradex, Atragranz, Atranex, Atranex 80WP, Atraphyt, Atrasan, Atrataf, Atrazerba FL, Atrazin, Atrazine, Azoprim, Azoprim 50, CET, Chromozin, Cyazin, Farmozine, G 30027, Gesamprim, Gesaprim, Gesaprim 50, Gesaprim 500, Gesaprim L, Herbatoxol, Hungazin, Hungazin PK, NSC 163046, Nortox 500SC, Nu-Trazine, Oleogesaprim, Oleogesaprim 200, Primatol A, Primoleo, Radazin, Radazin T, Triazine A 1294, Wonuk, Zealin L, Zeapos, Zeazin, Zeazine, Zeopos |
| InChI Formula | InChI=1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14) |
| SMILES | CCNc1nc(Cl)nc(NC(C)C)n1 |
| IUPAC Name | 6-chloro-4-N-ethyl-2-N-propan-2-yl-1,3,5-triazine-2,4-diamine |
| Show More |