missing translation for 'onlineSavingsMsg'
Learn More

ATR Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody

213.00€ - 507.00€

Specifications

Antigen ATR
Dilution Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30231590
View Documents
Novus Biologicals
NBP3-33305-20ul
20 μL
213.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30229217
View Documents
Novus Biologicals
NBP3-33305-100ul
100 μL
507.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ATR Monoclonal antibody specifically detects ATR in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications

Specifications

ATR
ELISA, Western Blot
Unconjugated
Rabbit
Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Immunology, p53 Pathway, Virology Bacteria and Parasites
PBS (pH 7.3), 50% glycerol, 0.05% BSA
545
IgG
Affinity purified
Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL
Monoclonal
Purified
RUO
Human, Mouse
ataxia telangiectasia and Rad3 related, Ataxia telangiectasia and Rad3-related protein, EC 2.7.11.1, FRAP-related protein 1, FRP1FRAP-related protein-1, MEC1, protein kinase ATR, Rad3 related protein, SCKL1, SCKLMEC1, mitosis entry checkpoint 1, homolog, serine/threonine-protein kinase ATR
A synthetic peptide corresponding to a sequence within amino acids 2100-2200 of human ATR (NP_001175.2).,, Sequence:, KAYEWEKAGRSDRVQMRNDLGKINKVITEHTNYLAPYQFLTAFSQLISRICHSHDEVFVVLMEIIAKVFLAYPQQAMWMMTAVSKSSYPMRVNRCKEILNK
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.