missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATPBD4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23993-1-AP-150UL
This item is not returnable.
View return policy
Description
Amidase that catalyzes the last step of diphthamide biosynthesis using ammonium and ATP. Diphthamide biosynthesis consists in the conversion of an L-histidine residue in the translation elongation factor (EEF2) to diphthamide.Specifications
| ATPBD4 | |
| Polyclonal | |
| Unconjugated | |
| DPH6 | |
| ATP binding domain 4, ATPBD4, Diphthamide synthase, Diphthamide synthetase, Diphthine-ammonia ligase, DPH6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66632, 89978 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7L8W6, Q9CQ28 | |
| DPH6 | |
| ATPBD4 Fusion Protein Ag21178 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction