missing translation for 'onlineSavingsMsg'
Learn More

ATP6V0D2 Monoclonal Antibody (OTI2D6), TrueMAB™, OriGene

Product Code. 17546494
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17546494 30 μL 30µL
17526494 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17546494 Supplier Origene Technologies Supplier No. TA812166S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ATP6V0D2 Monoclonal antibody specifically detects ATP6V0D2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ATP6V0D2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2D6
Concentration 0.5 to 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene LILRB3
Gene Accession No. O75022
Gene Alias 1620401A02Rik; AI324824; ATP6D2; Atp6v0d2; ATPase H+ transporting V0 subunit d2; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d2; ATPase, H+ transporting, lysosomal V0 subunit D2; ATPase, H+ transporting, V0 subunit D, isoform 2; FLJ38708; osteoclast-specific vacuolar ATP synthase; similar to ATPase, H+ transporting, V0 subunit D isoform 2; vacuolar proton pump subunit d 2; vacuolar proton-translocating ATPase d subunit d2; V-ATPase; V-ATPase subunit d 2; VMA6; V-type proton ATPase subunit d 2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 465-631 of human LILRB3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11025
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.