missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V0A4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00€ - 462.00€
Specifications
| Antigen | ATP6V0A4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ATP6V0A4 Polyclonal antibody specifically detects ATP6V0A4 in Human, Mouse samples. It is validated for Western BlotSpecifications
| ATP6V0A4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 50617 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| A4, ATP6N2, ATP6V0, ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 1B, ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 2 (38kD), ATPase, H+ transporting, lysosomal V0 subunit a4, MGC130016, MGC130017, noncatalytic accessory protein 1B, RDRTA2, RTA1C, RTADR, STV1, vacuolar proton pump 116 kDa accessory subunit, vacuolar proton pump, subunit 2, vacuolar proton translocating ATPase 116 kDa subunit a kidney isoform, V-ATPase 116 kDa, VPH1, VPP2, V-type proton ATPase 116 kDa subunit a, V-type proton ATPase 116 kDa subunit a isoform 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ATP6V0A4 (NP_065683.2). MVSVFRSEEMCLSQLFLQVEAAYCCVAELGELGLVQFKDLNMNVNSFQRKFVNEVRRCESLERILRFLEDEMQNEIVVQLLEKSPLTPLPREMITLETVLEKLEGELQEANQNQQALKQSFLELTELKYLLKKTQDFFETETNLADDFFTEDTSGLLELKAVPAYMTGKLGFIAGVINRERMASFERLLWRICRGNVYLKFSEMDAPLEDPVTKEEIQKNIFIIFYQGEQLRQKIKKICDGFRATVYPCPEPAVERREMLESVNVRLEDLITVITQTESH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title