missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ATP1B3 (Human) Recombinant Protein

Human ATP1B3 full-length ORF ( AAH11835, 1 a.a. - 279 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000483-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102021

  • 528.00€ / 25µg

Estimated Shipment: 12-07-2023
to see stock.
Add to basket

Description

Description

  • Sequence: MTKNEKKSLNQSLAEWKLFIYNPTTGEFLGRTAKSWGLILLFYLVFYGFLAALFSFTMWVMLQTLNDEVPKYRDQIPSPGLMVFPKPVTALEYTFSRSDPTSYAGYIEDLKKFLKPYTLEEQKNLTVCPDGALFEQKGPVYVACQFPISLLQACSGMNDPDFGYSQGNPCILVKMNRIIGLKPEGVPRIDCVSKNEDIPNVAVYPHNGMIDLKYFPYYGKKLHVGYLQPLVAVQVSFAPNNTGKEVTVECKIDGSANLKSQDDRDKFLGRVMFKITARA
Specifications

Specifications

AAH11835
ATPase, Na+/K+ transporting, beta 3 polypeptide
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATP1B3
GST
483
Wheat germ expression system
25 μg
ATPB-3, CD298, FLJ29027
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers