missing translation for 'onlineSavingsMsg'
Learn More

ATP1A3 Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen ATP1A3
Concentration 1000 μg/mL
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30236619
View Documents
Proteintech
25727-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30238064
View Documents
Proteintech
25727-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The ATP1A3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ATP1A3. This antibody recognizes human, mouse, rat antigen. The ATP1A3 antibody has been validated for the following applications: WB, IHC, IF-P, IP, ELISA analysis.
Specifications

Specifications

ATP1A3
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, ELISA
Unconjugated
Rabbit
Human, Mouse, Rat
DYT12; MGC13276; RDP
IgG
Antigen affinity purification
Polyclonal Antibody
1000 μg/mL
Polyclonal
Liquid
RUO
478
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.