missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ATP synthase C mature Antibody (SR2374), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27687
This item is not returnable.
View return policy
Description
ATP synthase C mature Monoclonal antibody specifically detects ATP synthase C mature in Human,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| ATP synthase C mature | |
| Monoclonal | |
| Unconjugated | |
| 50 mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 516 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| SR2374 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunoprecipitation 1:20 | |
| ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C1 (subunit 9), ATP5A, ATP5G, ATPase protein 9, ATPase subunit 9, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9), isoform 1, mitochondrial, mitochondrial ATP synthase, subunit 9, mitochondrial ATP synthase, subunit C | |
| A synthetic peptide of human ATP synthase C mature (Uniprot #: P05496/Q06055/P48201) | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction