missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP synthase C mature Antibody (S02-0D4), Novus Biologicals™
Rabbit Monoclonal Antibody
444.00€ - 691.00€
Specifications
| Antigen | ATP synthase C mature |
|---|---|
| Clone | S02-0D4 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611784
|
Novus Biologicals
NBP3-20012-100ul |
100 μg |
691.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681775
|
Novus Biologicals
NBP3-20012-50ul |
50 μg |
444.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATP synthase C mature Monoclonal antibody specifically detects ATP synthase C mature in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| ATP synthase C mature | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 516 | |
| IgG | |
| Affinity purified |
| S02-0D4 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C1 (subunit 9), ATP5A, ATP5G, ATPase protein 9, ATPase subunit 9, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9), isoform 1, mitochondrial, mitochondrial ATP synthase, subunit 9, mitochondrial ATP synthase, subunit C | |
| A synthetic peptide of human ATP synthase C mature (Uniprot # P05496) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title