Learn More
Atorvastatin Isopropyl Ester, Mikromol™
Description
Atorvastatin Isopropyl Ester
Specifications
Specifications
| Chemical Name or Material | Atorvastatin Isopropyl Ester |
| CAS | 1035205-25-4 |
| Analyte or Component Names | Atorvastatin Isopropyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C36 H41 F N2 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | (betaR,deltaR)-2-(4-Fluorophenyl)-beta,delta-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic Acid 1-Methylethyl Ester |
| InChI Formula | InChI=1S/C36H41FN2O5/c1-23(2)34-33(36(43)38-28-13-9-6-10-14-28)32(25-11-7-5-8-12-25)35(26-15-17-27(37)18-16-26)39(34)20-19-29(40)21-30(41)22-31(42)44-24(3)4/h5-18,23-24,29-30,40-41H,19-22H2,1-4H3,(H,38,43)/t29-,30-/m1/s1 |
| SMILES | CC(C)OC(=O)C[C@H](O)C[C@H](O)CCn1c(C(C)C)c(C(=O)Nc2ccccc2)c(c3ccccc3)c1c4ccc(F)cc4 |
| IUPAC Name | propan-2-yl (3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate |
| Show More |