missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atlantic Blue™ Anti-Human CD42b (AK2), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech AB-65163-100TESTS
This item is not returnable.
View return policy
Description
The CD42b antibody from Proteintech is a mouse monoclonal antibody generated with human platelets of human CD42b. This antibody recognizes human antigen. The CD42b antibody has been validated for the following applications: FC analysis. This antibody is atlantic blue™.Specifications
| CD42b | |
| Monoclonal | |
| 5 μl/test | |
| BSS; CD42B; CD42b-alpha; GP1B; MGC34595 | |
| Human platelets | |
| 100 Tests | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 |
| Flow Cytometry | |
| AK2 | |
| Atlantic Blue | |
| Mouse | |
| Purified by protein-A affinity chromatography | |
| RUO | |
| 2811 | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction