missing translation for 'onlineSavingsMsg'
Learn More

ATG9B Antibody, FITC, Novus Biologicals™

Product Code. 18440011 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18440011 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18440011 Supplier Novus Biologicals Supplier No. NBP177169F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATG9B Polyclonal antibody specifically detects ATG9B in Human, Mouse, Rat samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG9B
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias APG9L2APG9-like 2, ATG9 autophagy related 9 homolog B (S. cerevisiae), autophagy-related protein 9B, endothelial nitric oxide synthase antisense, nitric oxide synthase 3 antisense, Nitric oxide synthase 3-overlapping antisense gene protein, NOS3ASautophagy 9-like 2 protein, Protein sONE, SONEFLJ14885
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ATG9B. The immunogen is located within amino acids 850 - 900 of ATG9B. Amino Acid Squence: CPSQPPSPDEEKPSWSSD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 285973
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.