missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
217.00€ - 504.00€
Specifications
| Antigen | ATG5 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1 - 2 μg/ml, Flow Cytometry 1 - 2 μg/ml, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 1 - 2 μg/ml, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) 1 - 2 μg/ml |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18608116
|
Novus Biologicals
NBP3-11775 |
0.1 mg |
504.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18691597
|
Novus Biologicals
NBP3-11775SS |
0.025 mg |
217.00€
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG5 Polyclonal antibody specifically detects ATG5 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular)Specifications
| ATG5 | |
| Western Blot 1 - 2 μg/ml, Flow Cytometry 1 - 2 μg/ml, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 1 - 2 μg/ml, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) 1 - 2 μg/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| APG5, APG5 (Autophagy 5, S. Cerevisiae)-Like, APG5 autophagy 5-like (S. cerevisiae), APG5L, APG5-LIKE, ASP, ASPAPG5-LIKE, ATG5 Autophagy Related 5 Homolog, ATG5 autophagy related 5 homolog (S. cerevisiae), Autophagy protein 5, Autophagy Related 5, HAPG5, SCAR25 | |
| A partial recombinant protein produced in E. coli made to an internal portion of the human ATG5 protein produce in (between amino acids 80-400) [UniProt Q9H1Y0]. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Ubiquitin Proteasome Pathway | |
| PBS | |
| 9474 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title