missing translation for 'onlineSavingsMsg'
Learn More

ATG4A Antibody (SR1759), Novus Biologicals™

Product Code. 18673604 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18673604 100 μg 100µL
18609923 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18673604 Supplier Novus Biologicals Supplier No. NBP321808100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ATG4A Monoclonal antibody specifically detects ATG4A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG4A
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone SR1759
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias APG4 autophagy 4 homolog A (S. cerevisiae), APG4A, ATG4 autophagy related 4 homolog A (S. cerevisiae), AUTL2, AUT-like 2 cysteine endopeptidase, AUT-like 2, cysteine endopeptidase (S. cerevisiae), autophagin 2, autophagin-2, cysteine endopeptidase, cysteine protease ATG4A, EC 3.4.22, EC 3.4.22.-, hAPG4A
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATG4A (Uniprot #: Q8WYN0)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 115201
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.