missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ataxin-3 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
213.00€ - 507.00€
Specifications
| Antigen | Ataxin-3 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
30230712
|
Novus Biologicals
NBP3-33328-100ul |
100 μL |
507.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227119
|
Novus Biologicals
NBP3-33328-20ul |
20 μL |
213.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ataxin-3 Monoclonal antibody specifically detects Ataxin-3 in Mouse,Rat samples. It is validated for ELISA,Western BlotSpécification
| Ataxin-3 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neurodegeneration, Neuroscience | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 4287 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| ataxin 3, ATX3AT3, EC 3.4.19.12, EC 3.4.22, Machado-Joseph disease protein 1, MJD1, MJDolivopontocerebellar ataxia3, autosomal dominant, ataxin 3), olivopontocerebellar ataxia 3, Spinocerebellar ataxia type 3 protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 190-361 of human Ataxin-3 (ATXN3) (NP_004984.2).,, Sequence:, KLIGEELAQLKEQRVHKTDLERVLEANDGSGMLDEDEEDLQRALALSRQEIDMEDEEADLRRAIQLSMQGSSRNISQDMTQTSGTNLTSEELRKRREAYFEKQQQKQQQQQQQQQQGDLSGQSSHPCERPATSSGALGSDLGDAMSEEDMLQAAVTMSLETVRNDLKTEGKK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit