missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTM Method D5501 Ethanol Standard Kit, Dr. Ehrenstorfer
Click to view available options
Quantity:
5 x 2 mL
Unit Size:
10mL
Specifications
Specifications
| Chemical Name or Material | 2,2,4-Trimethylpentane∣Ethanol∣Methanol∣n-Heptane |
| CAS | 540-84-1, 64-17-5, 67-56-1, 142-82-5 |
| Analyte or Component Names | 2,2,4-Trimethylpentane∣Ethanol∣Methanol∣n-Heptane |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Quantity | 5 x 2 mL |
| Concentration or Composition (by Analyte or Components) | 2,2,4-Trimethylpentane 695300 μg/mL∣Methanol 0.1(0.6) wt%∣n-Heptane 0.5(10) wt%∣Ethanol 20.0(99.4) wt% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction