missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D5501 96% Ethanol QC Check Mixture in Ethanol, Dr. Ehrenstorfer

Product Code. 30318130
Change view
Click to view available options
Quantity:
10 x 2 mL
Unit Size:
20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318130 10 x 2 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318130 Supplier Dr. Ehrenstorfer Supplier No. DREGH09000173EL
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethanol∣Methanol∣n-Heptane
CAS 64-17-5, 67-56-1, 142-82-5
Analyte or Component Names Ethanol∣Methanol∣n-Heptane
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Ethanol
Quantity 10 x 2 mL
Concentration or Composition (by Analyte or Components) Methanol 1000 μg/mL∣n-Heptane 40000 μg/mL∣Ethanol 1000000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.