Learn More
ASTM Method D5443 Hydrocarbon Test Mixture, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 1,2,3-Trimethylbenzene∣1,2,4,5-Tetramethylbenzene∣1,2,4-Trimethylbenzene∣1,2,4-Trimethylcyclohexane∣1,2-Dimethylcyclohexane∣1-Hexene∣2,2,4-Trimethylpentane∣2,3-Dimethylbutane∣4-methyl-1-hexene∣Benzene∣Cyclohexane∣Cyclopentane∣Ethylbenzene∣Methylcyclohexane∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Nonane∣n-Octane∣n-Pentane∣n-Tetradecane∣n-Undecane∣o-Xylene (1,2-Dimethylbenzene)∣Pentamethylbenzene∣Propylbenzene∣Toluene∣trans-Decahydronaphthalene |
| CAS | 526-73-8, 95-93-2, 95-63-6, 2234-75-5, 583-57-3, 592-41-6 |
| Analyte or Component Names | 1,2,3-Trimethylbenzene∣1,2,4,5-Tetramethylbenzene∣1,2,4-Trimethylbenzene∣1,2,4-Trimethylcyclohexane∣1,2-Dimethylcyclohexane∣1-Hexene∣2,2,4-Trimethylpentane∣2,3-Dimethylbutane∣4-methyl-1-hexene∣Benzene∣Cyclohexane∣Cyclopentane∣Ethylbenzene∣Methylcyclohexane∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Nonane∣n-Octane∣n-Pentane∣n-Tetradecane∣n-Undecane∣o-Xylene (1,2-Dimethylbenzene)∣Pentamethylbenzene∣Propylbenzene∣Toluene∣trans-Decahydronaphthalene |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Cyclopentane 1 %∣n-Pentane 1 %∣1-Hexene 1.5 %∣4-methyl-1-hexene 1.5 %∣2,3-Dimethylbutane 2 %∣Cyclohexane 2 %∣n-Hexane 2 %∣Toluene 2.232 %∣Benzene 2.25 %∣n-Dodecane 3.25 %∣n-Undecane 3.47 %∣n-Heptane 3.5 %∣o-Xylene (1,2-Dimethylbenzene) 4.218 %∣1,2,4-Trimethylcyclohexane 4.25 %∣Methylcyclohexane 4.25 %∣n-Decane 4.25 %∣trans-Decahydronaphthalene 4.25 %∣1,2,4-Trimethylbenzene 4.471 %∣Ethylbenzene 4.5 %∣n-Nonane 4.5 %∣n-Tetradecane 4.5 %∣1,2,3-Trimethylbenzene 5 %∣1,2,4,5-Tetramethylbenzene 5 %∣1,2-Dimethylcyclohexane 5 %∣2,2,4-Trimethylpentane 5 %∣n-Octane 5 %∣Pentamethylbenzene 5 %∣Propylbenzene 5 % |