missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTM Method D4815 Ethanol/RFA oxygenate free gasoline 10% Wt, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Ethanol∣Regular Unleaded Gasoline |
| CAS | 64-17-5, 8006-61-9 |
| Analyte or Component Names | Ethanol∣Regular Unleaded Gasoline |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Quantity | 5 x 20 mL |
| Concentration or Composition (by Analyte or Components) | Ethanol 100000 μg/mL∣Regular Unleaded Gasoline 900000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?