Learn More
ASTM™ D6584 Individual Stock and Internal Standards Kit, Honeywell Fluka™
Product Code. 15929711 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. 15929711
Brand: Honeywell Fluka R13921EA
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Packaging Kit
Physical Form Liquid
Industry Type Biofuels
Quantity 1 Ea.
Grade Analytical Standard