Learn More
Hecht Karl™ Assistent™ Polyethylene Spray Bottles with Adjustable Jet
Product Code. 10043981
Change view
Click to view available options
Capacity (Metric):
∼1000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10043981 ∼1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10043981 Supplier Hecht Karl™ Supplier No. 49119010
  • ONLINE EXCLUSIVE PRICE
    14.10€
    10.85€ / Pack of 1
    Save 3.25€
    23% Off
Estimated Shipment: 27-11-2026
Specifications
Capacity (Metric) ∼1000 mL
Material Polyethylene
Quantity 1 Each
Certifications/Compliance ISO 9001 Certified
Product Type Spray Bottle