missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ASRGL1 Recombinant Protein

Brand:  Abnova™ H00080150-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194783

  • 349.00€ / 10µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH64963
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
45.9
8
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASRGL1
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
80150
ASRGL1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
ALP/ALP1/FLJ22316
ASRGL1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers