missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Aspartate Aminotransferase Polyclonal antibody specifically detects Aspartate Aminotransferase in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Aspartate Aminotransferase |
| Applications | Sandwich ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Sandwich ELISA 3 ng/mL - 100 ng/mL |
| Formulation | 1x PBS, pH 7.4 |
| Gene Alias | aspartate aminotransferase, cytoplasmic, ASTQTL1, CASPAT, CCAT, Cysteine aminotransferase, cytoplasmic, Cysteine transaminase, cytoplasmic, EC 2.6.1.1, EC 2.6.1.3, GIG18, Glutamate oxaloacetate transaminase 1, glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1), GOT1, growth-inhibiting protein 18, Transaminase A |
| Quantity | 1 Set |
| Regulatory Status | RUO |
| Research Discipline | Lipid and Metabolism |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?