missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASL Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67692-1-IG-150UL
This item is not returnable.
View return policy
Description
The ASL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ASL. This antibody recognizes human, mouse, rat, pig antigen. The ASL antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| ASL | |
| Monoclonal | |
| 1000 μg/mL | |
| ASAL | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| 50 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 1G9H3 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 435 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction