missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASK1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76674-0.025mg
This item is not returnable.
View return policy
Description
ASK1 Polyclonal specifically detects ASK1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| ASK1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Apoptosis signal-regulating kinase 1, ASK-1, ASK1MEKK 5, EC 2.7.11, MAP/ERK kinase kinase 5, MAPK/ERK kinase kinase 5, MAPKKK5EC 2.7.11.25, MEK kinase 5, MEKK5apoptosis signal regulating kinase 1, mitogen-activated protein kinase kinase kinase 5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-20 μg/mL | |
| BAA12684 | |
| MAP3K5 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human ASK1. The immunogen is located within amino acids 1260 - 1310 of ASK1. Amino Acid Squence: IEEKDQEIKHLKLKSQ | |
| 0.025 mg | |
| Apoptosis, Protein Kinase | |
| 4217 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction