missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
280.00€ - 588.00€
Specifications
| Antigen | ASC1 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18490921
|
Novus Biologicals
NBP1-89656-25ul |
25 μL |
280.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18048875
|
Novus Biologicals
NBP1-89656 |
0.1 mL |
588.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ASC1 Polyclonal specifically detects ASC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ASC1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| activating signal cointegrator 1, ASC-1, HsT17391, thyroid hormone receptor interactor 4, thyroid receptor interacting protein 4, Thyroid receptor-interacting protein 4, TR-interacting protein 4, TRIP-4 | |
| TRIP4 | |
| IgG | |
| Affinity Purified | |
| Specificity of human ASC1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, DNA Repair, Tumor Suppressors | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 9325 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ILQRDSNKSQKLLKKLMSGVENSGKVDISTKDLLPHQELRIKSGLEKAIKHKDKLLEFDRTSIRRTQVIDDESDYFASDSNQWLSKLERETLQKREEELRELRHASRLSKKVTIDFAGRKILEEENSLA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title