missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASC/TMS1 Antibody (1C3D7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
234.00€ - 582.00€
Specifications
| Antigen | ASC/TMS1 |
|---|---|
| Clone | 1C3D7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18294242
|
Novus Biologicals
NBP2-61682 |
0.1 mL |
582.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18658788
|
Novus Biologicals
NBP2-61682-0.025ml |
0.025 mL |
234.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ASC/TMS1 Monoclonal specifically detects ASC/TMS1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ASC/TMS1 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity | |
| 29108 | |
| This ASC/TMS1 Antibody (1C3D7) was developed against purified recombinant fragment of human ASC/TMS1 (AA: 1-120) expressed in E. Coli. | |
| Primary | |
| 21.6 kDa |
| 1C3D7 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| ASCMGC10332, CARD5TMS, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYD and CARD domain containing, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS-1, TMS1apoptosis-associated speck-like protein containing a CARD | |
| PYCARD | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title