missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arylsulfatase G/ARSG Antibody [HRP], Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals AF4600H
This item is not returnable.
View return policy
Description
Arylsulfatase G/ARSG Polyclonal antibody specifically detects Arylsulfatase G/ARSG in Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Arylsulfatase G/ARSG | |
| Polyclonal | |
| PBS | |
| Sheep | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| HRP | |
| arylsulfatase G, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.4, KIAA1001ASG | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Arylsulfatase G/ARSG, Gly17-Val525, Accession # Q3TYD4 | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 22901 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction