missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CtBP1 Antibody [mFluor Violet 610 SE], Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals AF4639MFV610
This item is not returnable.
View return policy
Description
CtBP1 Polyclonal antibody specifically detects CtBP1 in Human samples. It is validated for Western Blot
Specifications
| CtBP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| mFluor Violet 610 SE | |
| BARS, brefeldin A-ribosylated substrate, CTBP, CtBP1, C-terminal binding protein 1, C-terminal-binding protein 1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.158, MGC104684 | |
| E. coli-derived recombinant human CtBP1, Met1-Leu440, Accession # Q13363 | |
| 0.1 mL | |
| Cell Cycle and Replication, Wnt Signaling Pathway | |
| 1487 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction