Learn More
Articaine EP Impurity A (as Hydrochloride), Mikromol™
Description
Articaine EP Impurity A (as Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | Acetamidoarticaine Hydrochloride |
| CAS | 2984442-47-7 |
| Analyte or Component Names | Acetamidoarticaine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H18 N2 O3 S . HCl |
| Certifications/Compliance | ISO 9001 |
| Synonym | Methyl 3-[[2-(Propylamino)acetyl]amino]-4-methylthiophene-2-carboxylate Hydrochloride (Acetamidoarticaine Hydrochloride), Articaine Hydrochloride Imp. A (EP), Articaine Imp. A (EP), Articaine USP Related Compound A, Articaine USP RC A, Methyl 3-[[2-(Propylamino)acetyl]amino]-4-methylthiophene-2-carboxylate Hydrochloride, Articaine Hydrochloride Imp. A (EP) as Hydrochloride |
| InChI Formula | InChI=1S/C12H18N2O3S.ClH/c1-4-5-13-6-9(15)14-10-8(2)7-18-11(10)12(16)17-3;/h7,13H,4-6H2,1-3H3,(H,14,15);1H |
| SMILES | Cl.CCCNCC(=O)Nc1c(C)csc1C(=O)OC |
| IUPAC Name | methyl 4-methyl-3-[[2-(propylamino)acetyl]amino]thiophene-2-carboxylate;hydrochloride |
| Show More |