missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ART5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38394-20ul
This item is not returnable.
View return policy
Description
ART5 Polyclonal antibody specifically detects ART5 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| ART5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| ADP-ribosyltransferase 5, EC 2.4.2.31, ecto-ADP-ribosyltransferase 5, MGC22848, Mono(ADP-ribosyl)transferase 5, NAD(P)(+)--arginine ADP-ribosyltransferase 5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 23-200 of human ART5 (NP_443750.2).,, Sequence:, VPILPLGLAPDTFDDTYVGCAEEMEEKAAPLLKEEMAHHALLRESWEAAQETWEDKRRGLTLPPGFKAQNGIAIMVYTNSSNTLYWELNQAVRTGGGSRELYMRHFPFKALHFYLIRALQLLRGSGGCSRGPGEVVFRGVGSLRFEPKRLGDSVRLGQFASSSLDKAVAHRFGNATLF | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 116969 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction