missing translation for 'onlineSavingsMsg'
Learn More

ART2.2/Art2b Antibody (Nika102), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18757390 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.125 mL
Unit Size:
125µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18757390 0.125 mL 125µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18757390 Supplier Novus Biologicals Supplier No. NB10065853AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

ART2.2/Art2b Monoclonal antibody specifically detects ART2.2/Art2b in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ART2.2/Art2b
Applications Flow Cytometry
Classification Monoclonal
Clone Nika102
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias ADP-ribosyltransferase 2b, Rt6, Rt-6, Rt6.2, Rt6-2
Host Species Rat
Immunogen pME.CD8LF-ART2.2
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11872
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.