Learn More
Applied Biosystems™ Array Card Calibration Kit
Shop All Applied Biosystems Products
Click to view available options
Quantity:
9 arrays
Unit Size:
Each
Description
- Kit is used to maintain real-time PCR instruments with an array card block
- The kit includes region of interest (ROI), background, and dye calibrations, as well as RNaseP verification
- Please see the instrument user guide for full instructions on use
Specifications
Specifications
| Calibrated Dye | Proprietary Mix |
| For Use With (Equipment) | ViiA™ 7 Dx System, QuantStudio™ Dx |
| Quantity | 9 arrays |
| Calibration Type | Spectral Calibration |