Learn More
Applied Biosystems™ Array Card Calibration Kit
Product Code. 15880393 Shop All Applied Biosystems Products
Click to view available options
Quantity:
9 arrays
Unit Size:
Each
Product Code. 15880393
Brand: Applied Biosystems™ 4432507
The Array Card Calibration Kit is used to maintain real-time PCR instruments with an array card block. The kit includes region of interest (ROI), background, and dye calibrations, as well as RNaseP verification.
  • Kit is used to maintain real-time PCR instruments with an array card block
  • The kit includes region of interest (ROI), background, and dye calibrations, as well as RNaseP verification
  • Please see the instrument user guide for full instructions on use
TRUSTED_SUSTAINABILITY
Specifications
Calibrated Dye Proprietary Mix
For Use With (Equipment) ViiA™ 7 Dx System, QuantStudio™ Dx
Quantity 9 arrays
Calibration Type Spectral Calibration