Learn More
Arprinocid, TRC
Product Code. 17994535
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994535 50 mg 50mg
17931295 100 mg 100mg
17963105 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17994535 Supplier Toronto Research Chemicals Supplier No. TRCA73550050MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55779-18-5
Molecular Formula C12 H9 Cl F N5
Molecular Weight (g/mol) 277.68
Synonym 9-[(2-Chloro-6-fluorophenyl)methyl]-9H-purin-6-amine, 6-Amino-9-(2-chloro-6-fluorobenzyl)purine, Arpocox, Arprinocid, MK 302
IUPAC Name 9-[(2-chloro-6-fluorophenyl)methyl]purin-6-amine
SMILES Nc1ncnc2c1ncn2Cc3c(F)cccc3Cl
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C12H9ClFN5/c13-8-2-1-3-9(14)7(8)4-19-6-18-10-11(15)16-5-17-12(10)19/h1-3,5-6H,4H2,(H2,15,16,17)
Formula Weight 277.0531
Chemical Name or Material Arprinocid