missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ARP10 Polyclonal specifically detects ARP10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | ARP10 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | Q6NTF7 |
| Gene Alias | APOBEC-related protein 10, apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3H, ARP-10, ARP10Apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3H, DNA dC->dU-editing enzyme APOBEC-3H, EC 3.5.4.- |
| Gene Symbols | APOBEC3H |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?