Learn More
Aroclor 1254 1000 μg/mL in Isopropanol, Dr. Ehrenstorfer
Product Code. 30314608
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314608 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30314608 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010306IP
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Isopropanol